Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTLC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018327.2 | NP_060797.2 | 1, 2 | 405 | ||
XM_011529279.1 | XP_011527581.1 | 1, 2 | 173 | |||
XM_017027922.1 | XP_016883411.1 | 1, 2 | 173 | |||
GenBank mRNA | AK001974.1 | - | 1, 2 | 185 | ||
AK300184.1 | - | 1, 2 | 211 | |||
BC005205.1 | - | 1, 2 | 405 | |||
BC020656.1 | - | 1, 2 | 121 | |||
BC150644.1 | - | 1, 2 | 364 | 9057014 | ||
CF994231.1 | - | 1, 2 | 179 |