Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304748.1 | NP_001291677.1 | 3 | 730 | ||
NM_001304749.1 | NP_001291678.1 | 2 | 474 | |||
NM_018354.2 | NP_060824.1 | 2 | 881 | |||
XM_011529281.2 | XP_011527583.1 | 3 | 1922 | |||
XM_011529282.2 | XP_011527584.1 | 3 | 990 | |||
XM_011529283.2 | XP_011527585.1 | 3 | 1101 | |||
XM_011529284.2 | XP_011527586.1 | 3 | 1062 | |||
XM_011529285.2 | XP_011527587.1 | 2 | 247 | |||
XM_017027923.1 | XP_016883412.1 | 3 | 535 | |||
XM_017027924.1 | XP_016883413.1 | 3 | 495 | |||
XM_017027925.1 | XP_016883414.1 | 3 | 1111 | |||
GenBank mRNA | AK002052.1 | - | 2 | 881 | ||
AK126837.1 | - | 1 | 1467 | |||
AW628999.1 | - | 2 | 474 |