Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM149B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173348.1 | NP_775483.1 | 4, 5 | 649 | ||
XM_005269744.2 | XP_005269801.1 | 4, 5 | 649 | |||
XM_005269745.2 | XP_005269802.1 | 4, 5 | 649 | |||
XM_005269747.3 | XP_005269804.1 | 4, 5 | 424 | |||
XM_011539737.2 | XP_011538039.1 | 4, 5 | 452 | |||
XM_011539740.2 | XP_011538042.1 | 4, 5 | 649 | |||
XM_017016164.1 | XP_016871653.1 | 4, 5 | 453 | |||
XM_017016165.1 | XP_016871654.1 | 3, 4 | 296 | |||
XM_017016166.1 | XP_016871655.1 | 3, 4 | 317 | |||
XM_017016167.1 | XP_016871656.1 | 4, 5 | 649 | |||
GenBank mRNA | AB023191.1 | - | 3, 4 | 377 | ||
AK303436.1 | - | 4, 5 | 413 | |||
BC015394.1 | - | 2, 3 | 250 | 4431919 |