Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286349.1 | NP_001273278.1 | 9 | 1399 | ||
NM_018240.6 | NP_060710.3 | 11 | 1699 | |||
XM_005245305.4 | XP_005245362.1 | 11 | 1404 | |||
GenBank mRNA | AK001707.1 | - | 4 | 391 | ||
AK022708.1 | - | 7 | 749 | |||
AK297788.1 | - | 9 | 1027 | |||
AY017369.1 | - | 11 | 1295 | |||
AY302131.1 | - | 9 | 1394 | |||
BC025268.1 | - | 6 | 595 | |||
BC051356.1 | - | 4 | 362 | |||
BC067097.1 | - | 4 | 362 | |||
BC082969.1 | - | 4 | 362 | |||
BC109192.1 | - | 11 | 1362 | 40001501 | ||
BC109193.1 | - | 11 | 1362 | 40001502 |