Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242761.1 | NP_001229690.1 | 2, 3 | 377 | ||
NM_004746.3 | NP_004737.2 | 2, 3 | 377 | |||
XM_005258171.2 | XP_005258228.1 | 2, 3 | 418 | |||
XM_005258172.1 | XP_005258229.1 | 2, 3 | 413 | |||
XM_011525770.2 | XP_011524072.1 | 2, 3 | 418 | |||
XM_011525771.2 | XP_011524073.1 | 2, 3 | 418 | |||
GenBank mRNA | AK304101.1 | - | 2, 3 | 359 | ||
AK316099.1 | - | 2, 3 | 377 | |||
BC040718.1 | - | 2, 3 | 245 |