Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540885.1 | XP_011539187.1 | 1 | 338 | ||
XM_011540886.1 | XP_011539188.1 | 1 | 338 | |||
XM_011540888.2 | XP_011539190.1 | 1 | 338 | |||
XM_017000533.1 | XP_016856022.1 | 1 | 338 |