Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018268.3 | NP_060738.2 | 6, 7 | 851 | ||
XM_005248552.3 | XP_005248609.1 | 6, 7 | 818 | |||
XM_011543505.2 | XP_011541807.1 | 6, 7 | 613 | |||
XM_011543507.2 | XP_011541809.1 | 6, 7 | 818 | |||
XM_017009606.1 | XP_016865095.1 | 6, 7 | 1261 | |||
GenBank mRNA | AF115511.1 | - | 6, 7 | 600 | ||
AK001766.1 | - | 6, 7 | 797 | |||
AK021855.1 | - | 6, 7 | 637 | |||
AK023977.1 | - | 6, 7 | 553 | |||
AK300058.1 | - | 4, 5 | 462 | |||
BC040241.1 | - | 6, 7 | 603 | 3447269 | ||
KU178690.1 | - | 6, 7 | 531 |