Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRABD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194986.1 | NP_001181915.1 | 4 | 1063 | ||
XM_006710640.3 | XP_006710703.1 | 4 | 1539 | |||
XM_011541443.2 | XP_011539745.1 | 4 | 1539 | |||
XM_011541444.2 | XP_011539746.1 | 4 | 1540 | |||
XM_017001260.1 | XP_016856749.1 | 4 | 1539 | |||
GenBank mRNA | JQ653416.1 | - | 4 | 962 |