Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304745.1 | NP_001291674.1 | 2 | 483 | ||
NM_018283.3 | NP_060753.1 | 2 | 483 | |||
NR_136687.1 | 2 | 483 | ||||
NR_136688.1 | 2 | 483 | ||||
GenBank mRNA | AK001818.1 | - | 2 | 483 | ||
BC050698.1 | - | 2 | 311 | |||
BC064607.1 | - | 2 | 302 | |||
BC107875.1 | - | 2 | 356 | 6451308 | ||
BC133015.1 | - | 2 | 424 | 40147037 | ||
BC133017.1 | - | 2 | 424 | 40147039 | ||
DA000395.1 | - | 2 | 268 | |||
DA760263.1 | - | 2 | 289 |