Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287444.1 | NP_001274373.1 | 7, 8 | 1024 | ||
XM_006711894.3 | XP_006711957.1 | 7, 8 | 1018 | |||
XM_017004836.1 | XP_016860325.1 | 7, 8 | 1018 | |||
XM_017004837.1 | XP_016860326.1 | 7, 8 | 1018 | |||
XM_017004838.1 | XP_016860327.1 | 6, 7 | 889 | |||
GenBank mRNA | DY654549.1 | - | 3, 4 | 288 |