Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080393.1 | NP_001073862.1 | 6, 7 | 1146 | ||
XM_011534067.2 | XP_011532369.1 | 6, 7 | 1012 | |||
XM_011534068.1 | XP_011532370.1 | 6, 7 | 1031 | |||
XM_017007123.1 | XP_016862612.1 | 5, 6 | 763 | |||
XM_017007124.1 | XP_016862613.1 | 5, 6 | 892 | |||
GenBank mRNA | BC127733.1 | - | 5, 6 | 792 | 40127813 |