Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284406.1 | NP_001271335.1 | 5 | 556 | ||
NM_001284407.1 | NP_001271336.1 | 4 | 378 | |||
NM_001284408.1 | NP_001271337.1 | 3 | 240 | |||
NM_018306.3 | NP_060776.2 | 5 | 468 | |||
XM_011533937.2 | XP_011532239.1 | 6 | 723 | |||
GenBank mRNA | AK001898.1 | - | 4 | 310 | ||
AK092470.1 | - | 5 | 468 | |||
AK301984.1 | - | 5 | 556 | |||
BC020658.1 | - | 4 | 328 | 4768606 | ||
BG335433.1 | - | 3 | 179 |