Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145432.1 | NP_001138904.1 | 3 | 426 | ||
GenBank mRNA | BC006003.1 | - | 3 | 370 | ||
BC032431.1 | - | 3 | 425 | |||
BC040154.1 | - | 3 | 331 | |||
BC046171.2 | - | 3 | 373 | |||
BC062747.1 | - | 3 | 356 | |||
BC086307.1 | - | 3 | 389 | |||
DA733575.1 | - | 3 | 426 |