Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAYSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304793.1 | NP_001291722.1 | 3 | 4930 | ||
NM_018322.2 | NP_060792.1 | 2 | 523 | |||
GenBank mRNA | AK001963.1 | - | 2 | 426 | ||
AK124992.1 | - | 2 | 3184 | |||
AL136839.1 | - | 2 | 478 | |||
BC022007.2 | - | 2 | 447 | 4825803 | ||
CR627244.1 | - | 3 | 4814 | |||
DA977455.1 | - | 2 | 523 |