Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIMAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018326.2 | NP_060796.1 | 3 | 214 | ||
XM_005250017.1 | XP_005250074.1 | 3 | 230 | |||
GenBank mRNA | AF117333.1 | - | 3 | 182 | ||
AK001972.1 | - | 3 | 176 | |||
AK301441.1 | - | 3 | 233 | |||
AK301606.1 | - | 3 | 186 | |||
AW836736.1 | - | 259 | ||||
BC020657.1 | - | 3 | 191 | 4701647 | ||
BX476689.1 | - | 3 | 257 |