Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128592.1 | NP_001122064.1 | 3, 4 | 470 | ||
XM_011514591.2 | XP_011512893.1 | 3, 4 | 905 | |||
GenBank mRNA | BM738331.1 | - | 3, 4 | 447 |