Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYATL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010904.1 | NP_001010904.1 | 4 | 361 | ||
GenBank mRNA | DB246049.1 | - | 4 | 361 |