Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517548.2 | XP_011515850.1 | 1 | 86 | ||
XM_017013579.1 | XP_016869068.1 | 1 | 86 | |||
XM_017013581.1 | XP_016869070.1 | 1 | 86 | |||
XM_017013582.1 | XP_016869071.1 | 1 | 86 | |||
GenBank mRNA | DA237404.1 | - | 1 | 113 |