Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC642943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111003.1 | 2, 3 | 312 | |||
GenBank mRNA | BX097312.1 | - | 2, 3 | 296 | ||
DB062721.1 | - | 2, 3 | 312 | |||
HY033202.1 | - | 2, 3 | 308 |
Gene Symbol: ANKRD19P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028712.1 | - | 10, 11 | 2159 |