Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138459.3 | NP_612468.1 | 5 | 2952 | ||
GenBank mRNA | AJ420576.1 | - | 1 | 1221 | ||
AK025467.1 | - | 1 | 1183 | |||
AK125119.1 | - | 5 | 2914 | |||
AK126335.1 | - | 6 | 2665 | |||
BC110325.1 | - | 5 | 2907 | 5553827 |