Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110456.1 | 3 | 269 | |||
GenBank mRNA | BX353482.2 | - | 3 | 269 | ||
EF565109.1 | - | 2 | 167 |