Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018256.3 | NP_060726.3 | 11 | 1839 | ||
XM_011511469.2 | XP_011509771.1 | 10 | 1716 | |||
GenBank mRNA | AF242546.1 | - | 11 | 1222 | ||
AK001743.1 | - | 11 | 1276 | |||
AK022781.1 | - | 11 | 1284 | |||
AK022782.1 | - | 11 | 1284 | |||
AK056092.1 | - | 11 | 1839 | |||
AK299489.1 | - | 8 | 809 | |||
BC008082.2 | - | 11 | 1219 | 3050219 | ||
EF011617.1 | - | 11 | 1218 |