Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018266.2 | NP_060736.1 | 9 | 2032 | ||
NR_073506.1 | 10 | 2141 | ||||
XM_005247578.1 | XP_005247635.1 | 8 | 1876 | |||
XM_006713687.1 | XP_006713750.1 | 9 | 1940 | |||
XM_017006788.1 | XP_016862277.1 | 8 | 1913 | |||
GenBank mRNA | AK001764.1 | - | 9 | 1974 | ||
AK223248.1 | - | 9 | 1964 | |||
BC003192.1 | - | 10 | 2056 | |||
BC010070.2 | - | 10 | 2056 | |||
BC021277.2 | - | 9 | 1950 | 4857987 |