Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA17P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003574.1 | 12, 13 | 2363 | |||
GenBank mRNA | DQ266102.1 | - | 12, 13 | 2363 |