Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR42E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023977.1 | XP_016879466.1 | 9 | 1807 | ||
XM_017023978.1 | XP_016879467.1 | 8 | 2568 | |||
XM_017023979.1 | XP_016879468.1 | 8 | 2568 | |||
XM_017023980.1 | XP_016879469.1 | 8 | 2568 |