Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303538.1 | NP_001290467.1 | 3, 4 | 784 | ||
NM_001303539.1 | NP_001290468.1 | 3, 4 | 694 | |||
NM_001303540.1 | NP_001290469.1 | 2, 3 | 589 | |||
NM_018273.3 | NP_060743.2 | 4, 5 | 889 | |||
NR_130317.1 | 2, 3 | 543 | ||||
GenBank mRNA | AK129801.1 | - | 4, 5 | 592 | ||
AK291521.1 | - | 2, 3 | 290 | |||
AK294408.1 | - | 2, 3 | 290 | |||
AK297591.1 | - | 2, 3 | 244 | |||
AK297610.1 | - | 3, 4 | 485 | |||
AK298317.1 | - | 3, 4 | 395 | |||
AK303635.1 | - | 3, 4 | 397 | |||
AY358161.1 | - | 2, 3 | 281 | |||
BC016919.1 | - | 4, 5 | 581 | 4400601 | ||
BC040021.1 | - | 4, 5 | 587 |