Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001481.2 | NP_001001481.2 | 7 | 1324 | ||
NM_018299.4 | NP_060769.4 | 6 | 1291 | |||
NR_073119.1 | 6 | 1312 | ||||
NR_073120.1 | 6 | 1272 | ||||
NR_073121.1 | 5 | 1188 | ||||
GenBank mRNA | AK001873.1 | - | 6 | 1206 | ||
AK021735.1 | - | 6 | 1206 | |||
AK024050.1 | - | 7 | 1288 | |||
AK225554.1 | - | 6 | 1222 | |||
AK295792.1 | - | 6 | 1291 | |||
AY948289.1 | - | 7 | 1260 | |||
BC016326.1 | - | 6 | 1245 | 4093131 | ||
BX538145.1 | - | 5 | 1111 | |||
BX648427.1 | - | 5 | 1117 |