Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008744.1 | NP_001008744.1 | 2 | 257 | ||
NM_018319.3 | NP_060789.2 | 3 | 480 | |||
XM_005267847.2 | XP_005267904.1 | 3 | 1154 | |||
XM_005267848.2 | XP_005267905.1 | 2 | 498 | |||
XM_006720197.3 | XP_006720260.1 | 3 | 721 | |||
XM_006720198.3 | XP_006720261.1 | 2 | 376 | |||
XM_011536942.2 | XP_011535244.1 | 2 | 897 | |||
XM_011536943.2 | XP_011535245.1 | 2 | 290 | |||
XM_011536944.1 | XP_011535246.1 | 3 | 481 | |||
XM_017021439.1 | XP_016876928.1 | 3 | 388 | |||
XM_017021440.1 | XP_016876929.1 | 2 | 280 | |||
XM_017021441.1 | XP_016876930.1 | 2 | 469 | |||
XM_017021442.1 | XP_016876931.1 | 1 | 141 | |||
XM_017021445.1 | XP_016876934.1 | 1 | 141 | |||
GenBank mRNA | AF182002.1 | - | 1 | 166 | ||
AK001952.1 | - | 2 | 221 | |||
AK093235.1 | - | 3 | 1153 | |||
AK315549.1 | - | 2 | 205 | |||
BC006083.1 | - | 3 | 424 | |||
BC015474.1 | - | 3 | 431 | 3900062 | ||
BC107877.1 | - | 3 | 484 | 6597752 | ||
BU849335.1 | - | 3 | 483 | |||
BX161451.1 | - | 3 | 427 |