Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018358.2 | NP_060828.2 | 7 | 867 | ||
XM_011512959.2 | XP_011511261.1 | 7 | 871 | |||
GenBank mRNA | AK002060.1 | - | 7 | 844 | ||
AK024758.1 | - | 1 | 66 | |||
AK092415.1 | - | 7 | 750 | |||
AK097073.1 | - | 5 | 1127 | |||
AK290106.1 | - | 7 | 739 | |||
AK299256.1 | - | 6 | 845 | |||
AK299433.1 | - | 7 | 631 | |||
BC009253.2 | - | 7 | 746 | 3028775 | ||
BC035652.1 | - | 7 | 741 | |||
BC051754.2 | - | 7 | 741 | 6064628 | ||
BC051884.1 | - | 7 | 741 | 5105761 | ||
KU178694.1 | - | 7 | 734 | |||
KU178695.1 | - | 7 | 734 |