Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018360.2 | NP_060830.2 | 2 | 266 | ||
GenBank mRNA | AK002071.1 | - | 2 | 261 | ||
BC101572.1 | - | 2 | 233 | 8069078 | ||
BC101576.1 | - | 2 | 233 | 8069082 | ||
DB098819.1 | - | 2 | 266 |