Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024049.1 | 4 | 1138 | |||
NR_024050.1 | 3 | 496 | ||||
NR_131216.1 | 2 | 426 | ||||
NR_131217.1 | 3 | 711 | ||||
NR_131222.1 | 3 | 544 | ||||
GenBank mRNA | BG573316.1 | - | 3 | 712 | ||
BX107175.1 | - | 3 | 467 | |||
HE601934.1 | - | 1 | 280 |