Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF214454.1 | - | 6 | 480 | ||
AF327353.1 | - | 6 | 491 | |||
AK002100.1 | - | 7 | 586 | |||
AK293800.1 | - | 7 | 497 | |||
AK294978.1 | - | 5 | 346 | |||
AK295142.1 | - | 6 | 514 | |||
AK295327.1 | - | 7 | 610 | |||
AK308856.1 | - | 7 | 583 | |||
AK315000.1 | - | 6 | 492 | |||
BC063034.1 | - | 7 | 486 | |||
BC073853.1 | - | 6 | 443 | 3867010 | ||
BG702017.1 | - | 6 | 531 |