Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018368.3 | NP_060838.3 | 2, 3 | 530 | ||
XM_011535941.1 | XP_011534243.1 | 2, 3 | 380 | |||
GenBank mRNA | AF113224.1 | - | 2, 3 | 250 | ||
AF208863.1 | - | 1, 2 | 378 | |||
AK002102.1 | - | 2, 3 | 279 | |||
AK225187.1 | - | 2, 3 | 279 | |||
AK225537.1 | - | 2, 3 | 264 | |||
AK290069.1 | - | 2, 3 | 410 | |||
AY136817.1 | - | 1, 2 | 35 | |||
BC010360.1 | - | 2, 3 | 262 | |||
BC047073.1 | - | 2, 3 | 369 | 5274221 | ||
DA028310.1 | - | 2, 3 | 530 |