Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004822.2 | NP_004813.2 | 7 | 3556 | ||
GenBank mRNA | BC092429.1 | - | 1 | 656 |