Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018405.3 | NP_060875.2 | 2 | 248 | ||
XM_005258000.1 | XP_005258057.1 | 2 | 358 | |||
GenBank mRNA | AF407672.1 | - | 2 | 140 | ||
AJ272196.2 | - | 2 | 155 | |||
AK024772.1 | - | 2 | 168 | |||
BC012386.1 | - | 2 | 178 | 3882737 | ||
BC035483.1 | - | 2 | 232 | |||
DB472863.1 | - | 2 | 278 |