Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024768.2 | NP_079044.2 | 1 | 620 | ||
XM_005247781.3 | XP_005247838.1 | 1 | 645 | |||
XM_011513163.2 | XP_011511465.1 | 1 | 645 |