Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256574.1 | NP_001243503.1 | 2 | 1235 | ||
NM_001256575.1 | NP_001243504.1 | 2 | 1152 | |||
XM_011543696.2 | XP_011541998.1 | 2 | 1235 | |||
XM_011543697.2 | XP_011541999.1 | 2 | 1152 | |||
GenBank mRNA | DA032687.1 | - | 2 | 454 | ||
DA498140.1 | - | 2 | 538 |