Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174164.1 | NP_001167635.1 | 6 | 2463 | ||
XM_005250342.3 | XP_005250399.1 | 5 | 2463 | |||
XM_005250343.3 | XP_005250400.1 | 5 | 2372 | |||
XM_006715985.3 | XP_006716048.1 | 5 | 2476 | |||
XM_017012205.1 | XP_016867694.1 | 6 | 3282 |