Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005354.2 | NP_001005354.1 | 4 | 803 | ||
NM_018457.3 | NP_060927.1 | 4 | 953 | |||
GenBank mRNA | AF217517.1 | - | 1 | 866 | ||
AK074398.1 | - | 1 | 849 | |||
BC001464.2 | - | 4 | 939 | |||
BC004512.2 | - | 1 | 546 | |||
BC014257.2 | - | 1 | 853 | 4765821 | ||
BC016064.1 | - | 1 | 850 | 4852862 | ||
BC021019.1 | - | 1 | 821 | |||
BC066943.1 | - | 1 | 852 | 4817871 | ||
BC071979.1 | - | 1 | 784 | |||
BX640962.1 | - | 2 | 691 |