Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288732.1 | NP_001275661.1 | 8 | 2970 | ||
NM_001288733.1 | NP_001275662.1 | 8 | 2962 | |||
NM_018469.4 | NP_060939.3 | 8 | 2991 | |||
XM_011524998.1 | XP_011523300.1 | 8 | 2980 | |||
XM_011524999.1 | XP_011523301.1 | 8 | 2904 | |||
XM_011525000.1 | XP_011523302.1 | 8 | 2961 | |||
XM_017024846.1 | XP_016880335.1 | 8 | 2940 | |||
GenBank mRNA | AB051525.2 | - | 7 | 2789 | ||
AF163260.1 | - | 6 | 1007 | |||
AF220182.1 | - | 4 | 525 | |||
AK000369.1 | - | 3 | 296 | |||
AK000758.1 | - | 7 | 1206 | |||
AK091156.1 | - | 7 | 1959 | |||
AK091214.1 | - | 8 | 2887 | |||
AK314316.1 | - | 3 | 241 | |||
AL832371.1 | - | 8 | 2870 | |||
AL834251.2 | - | 8 | 2881 | |||
BC008896.2 | - | 7 | 2147 | |||
BC033661.1 | - | 5 | 673 | |||
BC036672.1 | - | 8 | 2941 | |||
BC040521.1 | - | 8 | 2895 | 4837394 | ||
BC040522.1 | - | 8 | 2943 | |||
BC108279.1 | - | 6 | 1028 | |||
CR627433.1 | - | 8 | 2963 |