Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005269920.3 | XP_005269977.1 | 7 | 1143 | ||
XM_006717903.3 | XP_006717966.1 | 8 | 2334 | |||
XM_006717905.3 | XP_006717968.1 | 7 | 2257 | |||
XM_011539876.2 | XP_011538178.1 | 6 | 2235 | |||
XM_011539877.2 | XP_011538179.1 | 9 | 2399 | |||
XM_011539878.2 | XP_011538180.1 | 7 | 2257 | |||
XM_011539879.2 | XP_011538181.1 | 6 | 2243 | |||
XM_011539883.2 | XP_011538185.1 | 6 | 1188 | |||
XM_017016349.1 | XP_016871838.1 | 11 | 2643 | |||
XM_017016350.1 | XP_016871839.1 | 8 | 2602 | |||
XM_017016351.1 | XP_016871840.1 | 7 | 2190 | |||
XM_017016352.1 | XP_016871841.1 | 7 | 2190 | |||
GenBank mRNA | AF131775.1 | - | 4 | 502 |