Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPSNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018376.3 | NP_060846.2 | 3 | 829 | ||
NR_130759.1 | 2 | 618 | ||||
NR_130760.1 | 3 | 829 | ||||
XM_011518839.1 | XP_011517141.1 | 3 | 829 | |||
XM_017014889.1 | XP_016870378.1 | 3 | 781 | |||
GenBank mRNA | AK002137.1 | - | 3 | 434 | ||
AK310397.1 | - | 2 | 174 | |||
BC005202.1 | - | 3 | 362 | 3933738 | ||
BC017914.1 | - | 3 | 400 | 4292375 | ||
BX647652.1 | - | 3 | 401 |