Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033834.1 | 4 | 2837 | |||
GenBank mRNA | AK056047.1 | - | 4 | 2092 | ||
AK056281.1 | - | 2 | 1296 | |||
AK123838.1 | - | 4 | 2837 |