Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146688.1 | NP_001140160.1 | 2, 3 | 252 | ||
NM_018433.5 | NP_060903.2 | 2, 3 | 521 | |||
XM_006712051.3 | XP_006712114.1 | 1, 2 | 224 | |||
XM_017004493.1 | XP_016859982.1 | 2, 3 | 540 | |||
XM_017004494.1 | XP_016859983.1 | 2, 3 | 310 | |||
GenBank mRNA | AB018285.2 | - | 2, 3 | 284 | ||
AL832150.1 | - | 2, 3 | 287 | |||
BC038297.2 | - | 2, 3 | 298 | |||
BG723267.1 | - | 2, 3 | 303 | |||
BX640698.1 | - | 2, 3 | 322 | |||
BX647271.1 | - | 2, 3 | 290 | |||
BX647752.1 | - | 2, 3 | 350 | |||
BX648841.1 | - | 2, 3 | 253 | |||
BX649109.1 | - | 2, 3 | 291 | |||
CR749581.1 | - | 2, 3 | 289 | |||
DB453517.1 | - | 2, 3 | 438 |