Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018448.4 | NP_060918.2 | 4 | 880 | ||
GenBank mRNA | AB020636.2 | - | 4 | 782 | ||
AK027783.1 | - | 1 | 89 | |||
AK074922.1 | - | 1 | 54 | |||
AK096010.1 | - | 4 | 786 | |||
AK292456.1 | - | 6 | 719 | |||
AK314358.1 | - | 4 | 768 | |||
AL136810.1 | - | 4 | 792 | |||
BC026220.2 | - | 2 | 231 | |||
BC050341.1 | - | 4 | 576 | 5265409 |