Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125749.1 | 2 | 642 | |||
NR_125750.1 | 3 | 557 | ||||
NR_125751.1 | 2 | 440 | ||||
GenBank mRNA | BM556191.1 | - | 2 | 642 |