Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146254.1 | NP_001139726.1 | 8 | 3132 | ||
NM_001146255.1 | NP_001139727.1 | 6 | 2865 | |||
NM_024825.3 | NP_079101.3 | 8 | 3156 | |||
XM_006722901.1 | XP_006722964.1 | 10 | 2328 | |||
XM_006722902.1 | XP_006722965.1 | 9 | 2193 | |||
XM_006722903.1 | XP_006722966.1 | 8 | 2061 | |||
XM_011528308.1 | XP_011526610.1 | 10 | 2362 | |||
XM_011528309.2 | XP_011526611.1 | 9 | 2287 | |||
XM_011528310.2 | XP_011526612.1 | 9 | 2250 | |||
XM_011528311.1 | XP_011526613.1 | 9 | 2229 | |||
XM_011528313.1 | XP_011526615.1 | 9 | 2297 | |||
XM_011528314.1 | XP_011526616.1 | 9 | 2218 | |||
XM_011528316.1 | XP_011526618.1 | 7 | 2030 |