Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITFG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018463.3 | NP_060933.3 | 4 | 410 | ||
NR_130744.1 | 4 | 409 | ||||
XM_005253707.4 | XP_005253764.1 | 4 | 379 | |||
GenBank mRNA | AF220048.1 | - | 664 | |||
AK027483.1 | - | 4 | 334 | |||
AK130319.1 | - | 3 | 940 | |||
AK291110.1 | - | 4 | 338 | |||
AK300905.1 | - | 4 | 338 | |||
AY599883.1 | - | 4 | 270 | |||
BC006552.2 | - | 4 | 295 | 3029909 | ||
BC013399.2 | - | 4 | 295 | 3030623 | ||
BQ058412.1 | - | 409 | ||||
KU178719.1 | - | 4 | 270 | |||
KU178720.1 | - | 4 | 270 | |||
KU178721.1 | - | 4 | 270 |