Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146310.1 | NP_001139782.1 | 5 | 713 | ||
NM_001282670.1 | NP_001269599.1 | 4 | 393 | |||
XM_011540914.2 | XP_011539216.1 | 5 | 398 | |||
XM_011540921.2 | XP_011539223.1 | 2 | 247 | |||
XM_017000553.1 | XP_016856042.1 | 3 | 245 | |||
GenBank mRNA | AK126870.1 | - | 4 | 393 | ||
AK127994.1 | - | 5 | 713 |