Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024628.1 | 4 | 487 | |||
NR_024629.1 | 3 | 324 | ||||
NR_027708.1 | 5 | 620 | ||||
NR_027709.1 | 5 | 704 | ||||
GenBank mRNA | AF220183.1 | - | 3 | 296 | ||
AK314921.1 | - | 3 | 324 | |||
BC104155.1 | - | 4 | 377 | |||
BC104156.1 | - | 4 | 377 | |||
BC112975.1 | - | 4 | 377 | |||
BC112976.1 | - | 4 | 377 |